The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000356.3 | NP_000347.2 | 11 - 12 | 1765 | 64 |
NM_001008657.2 | NP_001008657.1 | 12 - 13 | 1996 | 64 | |
NM_001135243.1 | NP_001128715.1 | 12 - 13 | 1996 | 64 | |
NM_001135244.1 | NP_001128716.1 | 12 - 13 | 1996 | 64 | |
NM_001135245.1 | NP_001128717.1 | 11 - 12 | 1765 | 64 | |
NM_001195141.1 | NP_001182070.1 | 12 - 13 | 1996 | 64 | |
XM_005268502.3 | XP_005268559.1 | 12 - 13 | 1933 | 64 | |
XM_005268503.3 | XP_005268560.1 | 12 - 13 | 1933 | 64 | |
XM_005268504.3 | XP_005268561.1 | 12 - 13 | 1933 | 64 | |
XM_005268505.3 | XP_005268562.1 | 12 - 13 | 1933 | 64 | |
XM_005268506.3 | XP_005268563.1 | 12 - 13 | 1933 | 64 | |
XM_005268507.3 | XP_005268564.1 | 11 - 12 | 1702 | 64 | |
XM_005268508.3 | XP_005268565.1 | 12 - 13 | 1933 | 64 | |
XM_005268509.3 | XP_005268566.1 | 12 - 13 | 1933 | 64 | |
XM_011537678.2 | XP_011535980.1 | 12 - 13 | 1933 | 64 | |
XM_017009792.1 | XP_016865281.1 | 12 - 13 | 1933 | 64 | |
XM_017009793.1 | XP_016865282.1 | 12 - 13 | 1933 | 64 | |
XM_017009794.1 | XP_016865283.1 | 12 - 13 | 1933 | 64 | |
XM_017009795.1 | XP_016865284.1 | 11 - 12 | 1702 | 64 | |
GenBank mRNA | AB209317.1 | - | 4 - 5 | 688 | 64 |
AK096467.1 | - | 8 - 9 | 1502 | 64 | |
AK225284.1 | - | 11 - 12 | 1690 | 64 | |
AK296918.1 | - | 5 - 6 | 712 | 64 | |
AK299167.1 | - | 5 - 6 | 953 | 64 | |
AK303611.1 | - | 12 - 13 | 1921 | 64 | |
AK307586.1 | - | 12 - 13 | 1921 | 64 | |
AY460334.1 | - | 12 - 13 | 1888 | 64 | |
BC008331.1 | - | 3 - 4 | 562 | 64 | |
BC011764.2 | - | 12 - 13 | 1917 | 64 | |
BC014559.2 | - | 12 - 13 | 1917 | 64 | |
BC016144.1 | - | 12 - 13 | 1916 | 64 | |
BC027252.1 | - | 11 - 12 | 1613 | 64 | |
BC033093.1 | - | 12 - 13 | 1917 | 64 | |
U40847.1 | - | 11 - 12 | 1750 | 64 | |
U76366.1 | - | 11 - 12 | 1732 | 64 |