The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000356.3 | NP_000347.2 | 6 - 7 | 744 | 71 |
NM_001135245.1 | NP_001128717.1 | 6 - 7 | 744 | 71 | |
XM_005268507.3 | XP_005268564.1 | 6 - 7 | 681 | 71 | |
XM_017009795.1 | XP_016865284.1 | 6 - 7 | 681 | 71 | |
GenBank mRNA | AK225284.1 | - | 6 - 7 | 669 | 71 |
BC027252.1 | - | 6 - 7 | 592 | 71 | |
U40847.1 | - | 6 - 7 | 729 | 71 | |
U76366.1 | - | 6 - 7 | 711 | 71 |