The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020764.3 | NP_065815.1 | 13 - 14 | 1354 | 150 |
GenBank mRNA | AB037727.1 | - | 5 - 6 | 491 | 150 |
AF451977.1 | - | 13 - 14 | 1420 | 150 | |
BC042011.1 | - | 4 - 5 | 150 |