The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020764.3 | NP_065815.1 | 9 - 10 | 966 | 93 |
GenBank mRNA | AB037727.1 | - | 1 - 2 | 103 | 93 |
AF451977.1 | - | 9 - 10 | 1032 | 93 |