The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000296.3 | NP_000287.3 | 45 - 46 | 12653 | 62 |
NM_001009944.2 | NP_001009944.2 | 45 - 46 | 12656 | 62 | |
XM_005255370.3 | XP_005255427.1 | 35 - 36 | 10243 | 62 | |
XM_011522528.2 | XP_011520830.1 | 46 - 47 | 12681 | 62 | |
XM_011522529.2 | XP_011520831.1 | 46 - 47 | 12679 | 62 | |
XM_011522530.2 | XP_011520832.1 | 46 - 47 | 12486 | 62 | |
XM_011522532.2 | XP_011520834.1 | 45 - 46 | 12414 | 62 | |
XM_011522534.2 | XP_011520836.1 | 46 - 47 | 12471 | 62 | |
XM_011522535.2 | XP_011520837.1 | 37 - 38 | 10703 | 62 | |
XM_011522537.2 | XP_011520839.1 | 35 - 36 | 9614 | 62 | |
XM_017023285.1 | XP_016878774.1 | 36 - 37 | 10258 | 62 | |
GenBank mRNA | AK309422.1 | - | 3 - 4 | 686 | 62 |
L33243.1 | - | 45 - 46 | 12655 | 62 | |
U24497.1 | - | 45 - 46 | 12582 | 62 |