Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PKD1
- Entrez Gene ID
-
5310
- Gene Name
- polycystin 1, transient receptor potential channel interacting
- Gene Aliases
-
PBP, Pc-1, TRPP1
- Chromosome Location
-
Chr.16: 2088708 - 2135898 on Build GRCh38
- Gene Symbol
- PKD1P1
- Entrez Gene ID
-
339044
- Gene Name
- polycystin 1, transient receptor potential channel interacting pseudogene 1
- Gene Aliases
-
HG1, NPIP
- Chromosome Location
-
Chr.16: 16317609 - 16350608 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_036447.1 |
|
5 - 6 |
782 |
60 |
GenBank mRNA |
AK304826.1 |
- |
5 - 6 |
782 |
60 |
View Full Product Details