Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LZTFL1
Entrez Gene ID
54585
Gene Name
leucine zipper transcription factor like 1
Gene Aliases
BBS17
Chromosome Location
Chr.3: 45823316 - 45916042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276378.1 NP_001263307.1 11 - 12 1325 140
NM_020347.3 NP_065080.1 9 - 10 1044 140
NR_075080.1 8 - 9 919 140
XM_011533838.2 XP_011532140.1 9 - 10 968 140
XM_017006644.1 XP_016862133.1 12 - 13 1682 140
XM_017006645.1 XP_016862134.1 11 - 12 1557 140
GenBank mRNA AJ297351.1 - 9 - 10 1001 140
AK093705.1 - 11 - 12 1643 140
BC025988.2 - 9 - 10 973 140
BC042483.1 - 9 - 10 1004 140
BX640604.1 - 9 - 10 1029 140

View Full Product Details