Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LZTFL1
Entrez Gene ID
54585
Gene Name
leucine zipper transcription factor like 1
Gene Aliases
BBS17
Chromosome Location
Chr.3: 45823316 - 45916042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006644.1 XP_016862133.1 3 - 4 718 73
XM_017006645.1 XP_016862134.1 3 - 4 718 73

View Full Product Details