Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NCKIPSD
- Entrez Gene ID
-
51517
- Gene Name
- NCK interacting protein with SH3 domain
- Gene Aliases
-
AF3P21, DIP, DIP1, ORF1, SPIN90, VIP54, WASLBP, WISH
- Chromosome Location
-
Chr.3: 48663814 - 48685941 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ242655.2 |
- |
12 - 13 |
2065 |
89 |
View Full Product Details