Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPNS1
Entrez Gene ID
83985
Gene Name
sphingolipid transporter 1 (putative)
Gene Aliases
HSpin1, LAT, PP2030, SPIN1, SPINL, nrs
Chromosome Location
Chr.16: 28974767 - 28984769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142448.1 NP_001135920.1 5 - 6 737 68
NM_001142449.1 NP_001135921.1 3 - 4 908 68
NM_001142450.1 NP_001135922.1 5 - 6 887 68
NM_001142451.1 NP_001135923.1 4 - 5 974 68
NM_032038.2 NP_114427.1 4 - 5 974 68
XM_006721096.3 XP_006721159.1 5 - 6 744 68
XM_017023757.1 XP_016879246.1 5 - 6 745 68
XM_017023758.1 XP_016879247.1 5 - 6 741 68
XM_017023759.1 XP_016879248.1 5 - 6 740 68
GenBank mRNA AF212371.1 - 4 - 5 921 68
AF370423.1 - 5 - 6 869 68
AK095677.1 - 4 - 5 971 68
AK289787.1 - 4 - 5 969 68
AL390215.1 - 3 - 4 870 68
AL524908.3 - 5 - 6 862 68
BC006156.1 - 4 - 5 585 68
BC008325.1 - 4 - 5 799 68
BC038961.1 - 5 - 6 751 68
BC047741.1 - 4 - 5 948 68
BC065235.1 - 5 - 6 719 68

View Full Product Details