The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142448.1 | NP_001135920.1 | 9 - 10 | 1289 | 67 |
NM_001142449.1 | NP_001135921.1 | 6 - 7 | 1304 | 67 | |
NM_001142450.1 | NP_001135922.1 | 9 - 10 | 1439 | 67 | |
NM_001142451.1 | NP_001135923.1 | 7 - 8 | 1370 | 67 | |
NM_032038.2 | NP_114427.1 | 8 - 9 | 1526 | 67 | |
XM_006721096.3 | XP_006721159.1 | 8 - 9 | 1140 | 67 | |
XM_017023757.1 | XP_016879246.1 | 9 - 10 | 1312 | 67 | |
XM_017023758.1 | XP_016879247.1 | 9 - 10 | 1308 | 67 | |
XM_017023759.1 | XP_016879248.1 | 9 - 10 | 1292 | 67 | |
GenBank mRNA | AF212371.1 | - | 8 - 9 | 1473 | 67 |
AF370423.1 | - | 9 - 10 | 1421 | 67 | |
AL390215.1 | - | 6 - 7 | 1266 | 67 | |
BC006156.1 | - | 8 - 9 | 1137 | 67 | |
BC008325.1 | - | 8 - 9 | 1351 | 67 | |
BC038961.1 | - | 9 - 10 | 1303 | 67 | |
BC047741.1 | - | 7 - 8 | 1344 | 67 | |
BC065235.1 | - | 9 - 10 | 1271 | 67 | |
CD368853.1 | - | 4 - 5 | 67 |