The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326411.1 | NP_001313340.1 | 4 - 5 | 913 | 66 |
NM_001326412.1 | NP_001313341.1 | 4 - 5 | 850 | 66 | |
NM_001326413.1 | NP_001313342.1 | 4 - 5 | 999 | 66 | |
NM_001326414.1 | NP_001313343.1 | 5 - 6 | 1079 | 66 | |
NM_001326415.1 | NP_001313344.1 | 5 - 6 | 1253 | 66 | |
NM_001326416.1 | NP_001313345.1 | 7 - 8 | 1442 | 66 | |
NM_001326417.1 | NP_001313346.1 | 6 - 7 | 1333 | 66 | |
NM_001326418.1 | NP_001313347.1 | 3 - 4 | 833 | 66 | |
NM_001326419.1 | NP_001313348.1 | 6 - 7 | 1333 | 66 | |
NM_001326420.1 | NP_001313349.1 | 3 - 4 | 833 | 66 | |
NM_001326421.1 | NP_001313350.1 | 4 - 5 | 913 | 66 | |
NM_014338.3 | NP_055153.1 | 5 - 6 | 885 | 66 | |
NM_178022.1 | NP_821141.1 | 4 - 5 | 794 | 66 | |
GenBank mRNA | AK298474.1 | - | 4 - 5 | 794 | 66 |
BC001482.2 | - | 5 - 6 | 885 | 66 | |
CR456540.1 | - | 4 - 5 | 563 | 66 |