Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 4 - 5 751 90
NM_001278630.1 NP_001265559.1 4 - 5 608 90
NM_152786.2 NP_689999.1 4 - 5 824 90
XM_011518477.1 XP_011516779.1 4 - 5 751 90
XM_011518478.1 XP_011516780.1 4 - 5 751 90
GenBank mRNA AK313867.1 - 4 - 5 740 90
BC026884.1 - 4 - 5 790 90
BC033037.2 - 4 - 5 709 90

View Full Product Details