The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256819.1 | NP_001243748.1 | 1 - 2 | 540 | 100 |
GenBank mRNA | BM666773.1 | - | 3 - 4 | 100 |