Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
Chromosome Location
Chr.1: 171485496 - 171593511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015172.3 NP_055987.2 16 - 17 5024 59
XM_005245015.2 XP_005245072.1 16 - 17 5061 59
XM_005245016.4 XP_005245073.1 16 - 17 5041 59
XM_005245017.2 XP_005245074.1 16 - 17 5061 59
XM_005245018.2 XP_005245075.1 16 - 17 5036 59
XM_005245019.2 XP_005245076.1 16 - 17 5061 59
XM_005245020.2 XP_005245077.1 16 - 17 5061 59
XM_005245023.2 XP_005245080.1 16 - 17 5061 59
XM_005245024.2 XP_005245081.1 16 - 17 5061 59
XM_005245025.2 XP_005245082.1 16 - 17 5061 59
XM_005245026.2 XP_005245083.1 16 - 17 5061 59
XM_005245027.2 XP_005245084.1 16 - 17 5061 59
XM_005245028.2 XP_005245085.1 16 - 17 5061 59
XM_005245029.2 XP_005245086.1 16 - 17 5061 59
XM_006711238.3 XP_006711301.1 16 - 17 5041 59
XM_017000760.1 XP_016856249.1 16 - 17 5026 59
XM_017000761.1 XP_016856250.1 16 - 17 5061 59
XM_017000762.1 XP_016856251.1 16 - 17 5061 59
GenBank mRNA AB029019.2 - 1 - 2 1970 59
AF488829.1 - 15 - 16 4758 59
AL096857.1 - 15 - 16 4815 59
AL162004.1 - 1 - 2 401 59

View Full Product Details