Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
Chromosome Location
Chr.1: 171485496 - 171593511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015172.3 NP_055987.2 8 - 9 1230 143
XM_005245015.2 XP_005245072.1 8 - 9 1267 143
XM_005245016.4 XP_005245073.1 8 - 9 1247 143
XM_005245017.2 XP_005245074.1 8 - 9 1267 143
XM_005245018.2 XP_005245075.1 8 - 9 1242 143
XM_005245019.2 XP_005245076.1 8 - 9 1267 143
XM_005245020.2 XP_005245077.1 8 - 9 1267 143
XM_005245023.2 XP_005245080.1 8 - 9 1267 143
XM_005245024.2 XP_005245081.1 8 - 9 1267 143
XM_005245025.2 XP_005245082.1 8 - 9 1267 143
XM_005245026.2 XP_005245083.1 8 - 9 1267 143
XM_005245027.2 XP_005245084.1 8 - 9 1267 143
XM_005245028.2 XP_005245085.1 8 - 9 1267 143
XM_005245029.2 XP_005245086.1 8 - 9 1267 143
XM_006711238.3 XP_006711301.1 8 - 9 1247 143
XM_017000760.1 XP_016856249.1 8 - 9 1232 143
XM_017000761.1 XP_016856250.1 8 - 9 1267 143
XM_017000762.1 XP_016856251.1 8 - 9 1267 143
GenBank mRNA AF253978.1 - 7 - 8 1035 143
AF488829.1 - 7 - 8 964 143
AK298887.1 - - 261 143
AL096857.1 - 7 - 8 1021 143
BC001260.1 - 8 - 9 1201 143
BC026108.1 - 8 - 9 1207 143
BC045713.1 - 8 - 9 1202 143
BC058930.1 - 8 - 9 1213 143
BC066968.1 - 8 - 9 1251 143

View Full Product Details