The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304331.1 | NP_001291260.1 | 12 - 13 | 1930 | 70 |
NM_001304332.1 | NP_001291261.1 | 12 - 13 | 1930 | 70 | |
XM_006711586.2 | XP_006711649.1 | 12 - 13 | 1960 | 70 | |
XM_017002606.1 | XP_016858095.1 | 12 - 13 | 1960 | 70 | |
XM_017002607.1 | XP_016858096.1 | 11 - 12 | 1825 | 70 | |
XM_017002608.1 | XP_016858097.1 | 12 - 13 | 1960 | 70 | |
XM_017002609.1 | XP_016858098.1 | 11 - 12 | 1825 | 70 | |
XM_017002610.1 | XP_016858099.1 | 12 - 13 | 1960 | 70 | |
GenBank mRNA | AK295757.1 | - | 6 - 7 | 658 | 70 |
EF428334.1 | - | 16 - 17 | 1875 | 70 |