Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KNOP1
Entrez Gene ID
400506
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
101F10.1, C16orf88, FAM191A, TSG118
Chromosome Location
Chr.16: 19706347 - 19718240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012991.2 NP_001013009.2 2 - 3 1003 95
XM_005255317.4 XP_005255374.1 4 - 5 1198 95
XM_006721046.3 XP_006721109.1 3 - 4 1361 95
XM_011545855.1 XP_011544157.1 3 - 4 1247 95
GenBank mRNA BC117562.1 - 2 - 3 1003 95

View Full Product Details