The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012991.2 | NP_001013009.2 | 3 - 4 | 1083 | 78 |
XM_005255317.4 | XP_005255374.1 | 5 - 6 | 1278 | 78 | |
XM_006721046.3 | XP_006721109.1 | 4 - 5 | 1441 | 78 | |
XM_011545855.1 | XP_011544157.1 | 4 - 5 | 1327 | 78 | |
XM_017023231.1 | XP_016878720.1 | 4 - 5 | 1219 | 78 | |
XM_017023232.1 | XP_016878721.1 | 3 - 4 | 1364 | 78 | |
GenBank mRNA | BC117562.1 | - | 3 - 4 | 1083 | 78 |