The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020660.2 | NP_065711.1 | 1 - 2 | 168 | 71 |
GenBank mRNA | BC069339.1 | - | 1 - 2 | 75 | 71 |
BC112110.1 | - | 1 - 2 | 168 | 71 | |
BC112114.1 | - | 1 - 2 | 168 | 71 | |
BC143788.1 | - | 1 - 2 | 168 | 71 |