Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBN1
Entrez Gene ID
29855
Gene Name
ubinuclein 1
Gene Aliases
VT, VT4
Chromosome Location
Chr.16: 4846665 - 4882401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079514.2 NP_001072982.1 6 - 7 1289 132
NM_001288656.1 NP_001275585.1 5 - 6 723 132
XM_005255277.3 XP_005255334.1 6 - 7 1474 132
XM_005255278.3 XP_005255335.1 6 - 7 2438 132
XM_005255279.3 XP_005255336.1 6 - 7 1474 132
XM_011522465.1 XP_011520767.1 6 - 7 1474 132
XM_011522466.2 XP_011520768.1 2 - 3 186 132
XM_011522467.1 XP_011520769.1 6 - 7 1474 132
XM_017023189.1 XP_016878678.1 6 - 7 1474 132
XM_017023190.1 XP_016878679.1 6 - 7 1474 132
XM_017023192.1 XP_016878681.1 6 - 7 1474 132
GenBank mRNA AF108460.1 - 6 - 7 784 132
AF108461.1 - 5 - 6 1371 132
AK300131.1 - 5 - 6 723 132
AW385352.1 - 3 - 4 302 132

View Full Product Details