Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGR2
Entrez Gene ID
10551
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
Chromosome Location
Chr.7: 16791811 - 16805114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006408.3 NP_006399.1 4 - 5 440 111
XM_005249581.4 XP_005249638.1 4 - 5 409 111
GenBank mRNA AF007791.1 - 3 - 4 263 111
AF038451.1 - 4 - 5 311 111
AF087879.1 - 4 - 5 402 111
AF088867.1 - 4 - 5 400 111
AF115926.1 - 4 - 5 310 111
AK223067.1 - 4 - 5 331 111
AY359009.1 - 4 - 5 400 111
BC015503.1 - 4 - 5 309 111
BT007048.1 - 3 - 4 253 111
FJ224307.1 - 3 - 4 263 111
KJ767789.1 - 4 - 5 414 111

View Full Product Details