Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SASH3
Entrez Gene ID
54440
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
753P9, CXorf9, HACS2, SH3D6C, SLY
Chromosome Location
Chr.X: 129779916 - 129795201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018990.3 NP_061863.1 4 - 5 619 93
XM_006724763.1 XP_006724826.1 4 - 5 586 93
GenBank mRNA AK292023.1 - 4 - 5 567 93
AK296665.1 - 4 - 5 521 93
AK314869.1 - 4 - 5 586 93
AL049683.1 - 4 - 5 544 93
BC051881.1 - 4 - 5 551 93

View Full Product Details