The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003399.5 | NP_003390.4 | 17 - 18 | 1848 | 60 |
GenBank mRNA | BC126174.1 | - | 17 - 18 | 1765 | 60 |
BC143901.1 | - | 17 - 18 | 1765 | 60 | |
BQ224207.1 | - | 4 - 5 | 289 | 60 | |
U90724.2 | - | 17 - 18 | 1868 | 60 |