The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039465.1 | NP_001034554.1 | 1 - 2 | 267 | 62 |
XM_017021593.1 | XP_016877082.1 | 1 - 2 | 237 | 62 | |
GenBank mRNA | BC018823.2 | - | 1 - 2 | 225 | 62 |
BU928782.1 | - | 1 - 2 | 270 | 62 | |
BX247988.1 | - | 1 - 2 | 207 | 62 | |
U30827.1 | - | 1 - 2 | 197 | 62 | |
U30884.1 | - | 1 - 2 | 197 | 62 |