The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006252.3 | NP_006243.2 | 8 - 9 | 1490 | 119 |
XM_017001692.1 | XP_016857181.1 | 8 - 9 | 2955 | 119 | |
XM_017001693.1 | XP_016857182.1 | 8 - 9 | 1575 | 119 | |
XM_017001694.1 | XP_016857183.1 | 9 - 10 | 1483 | 119 | |
GenBank mRNA | AK314011.1 | - | 8 - 9 | 1490 | 119 |
BC069680.1 | - | 8 - 9 | 1462 | 119 | |
BC069740.1 | - | 8 - 9 | 1463 | 119 | |
BC069823.1 | - | 8 - 9 | 1463 | 119 | |
EF056019.1 | - | 8 - 9 | 1419 | 119 | |
FJ200485.1 | - | 6 - 7 | 1183 | 119 | |
FJ200486.1 | - | 7 - 8 | 1325 | 119 | |
HQ628625.1 | - | 7 - 8 | 1277 | 119 | |
U06454.1 | - | 8 - 9 | 1483 | 119 |