Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
Chromosome Location
Chr.20: 20034361 - 20056046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278625.1 NP_001265554.1 5 - 6 938 67
NM_001278626.1 NP_001265555.1 5 - 6 751 67
NM_001278627.1 NP_001265556.1 5 - 6 661 67
NM_001278628.1 NP_001265557.1 4 - 5 609 67
NM_016652.5 NP_057736.4 5 - 6 974 67
GenBank mRNA AB016840.1 - 4 - 5 571 67
AF111802.1 - 4 - 5 565 67
AF255443.2 - 5 - 6 938 67
AF318302.1 - 4 - 5 556 67
AF318303.1 - 5 - 6 974 67
AF318304.1 - 5 - 6 751 67
AF318305.1 - 5 - 6 661 67
AK022908.1 - 3 - 4 286 67
AK023246.1 - 1 - 2 121 67
AK292799.1 - 4 - 5 479 67
AL832435.1 - 4 - 5 583 67
BI550198.1 - 4 - 5 603 67

View Full Product Details