Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SRSF11
- Entrez Gene ID
-
9295
- Gene Name
- serine and arginine rich splicing factor 11
- Gene Aliases
-
NET2, SFRS11, dJ677H15.2, p54
- Chromosome Location
-
Chr.1: 70205682 - 70252020 on Build GRCh38
View Full Product Details