The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001958.3 | NP_001949.1 | 5 - 6 | 1100 | 80 |
GenBank mRNA | AB209064.1 | - | 3 - 4 | 3340 | 80 |
AB451389.1 | - | 4 - 5 | 771 | 80 | |
AK307536.1 | - | 5 - 6 | 868 | 80 | |
BC000432.2 | - | 4 - 5 | 839 | 80 | |
BC018855.2 | - | 3 - 4 | 481 | 80 | |
BC110409.1 | - | 5 - 6 | 1069 | 80 | |
L10340.1 | - | 4 - 5 | 688 | 80 | |
X70940.1 | - | 4 - 5 | 854 | 80 |