Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REEP4
Entrez Gene ID
80346
Gene Name
receptor accessory protein 4
Gene Aliases
C8orf20, PP432, Yip2c
Chromosome Location
Chr.8: 22138014 - 22141951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316964.1 NP_001303893.1 4 - 5 777 80
NM_001316965.1 NP_001303894.1 4 - 5 777 80
NM_025232.3 NP_079508.2 4 - 5 777 80
GenBank mRNA AF218014.1 - 4 - 5 555 80
AK025899.1 - 4 - 5 707 80
AK025930.1 - 4 - 5 410 80
AK172846.1 - 4 - 5 712 80
AK302352.1 - 4 - 5 594 80
AY562242.1 - 4 - 5 308 80
BC013048.1 - 4 - 5 761 80
BC050622.1 - 4 - 5 679 80
BG695963.1 - 4 - 5 366 80
CR457353.1 - 4 - 5 308 80

View Full Product Details