Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REEP4
Entrez Gene ID
80346
Gene Name
receptor accessory protein 4
Gene Aliases
C8orf20, PP432, Yip2c
Chromosome Location
Chr.8: 22138014 - 22141951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316964.1 NP_001303893.1 5 - 6 884 122
NM_025232.3 NP_079508.2 5 - 6 884 122
GenBank mRNA AF218014.1 - 5 - 6 662 122
AK025899.1 - 5 - 6 814 122
AK025930.1 - 5 - 6 517 122
AK172846.1 - 5 - 6 819 122
AY562242.1 - 5 - 6 415 122
BC013048.1 - 5 - 6 868 122
BG695963.1 - 5 - 6 473 122
CR457353.1 - 5 - 6 415 122

View Full Product Details