Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REEP4
Entrez Gene ID
80346
Gene Name
receptor accessory protein 4
Gene Aliases
C8orf20, PP432, Yip2c
Chromosome Location
Chr.8: 22138014 - 22141951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025232.3 NP_079508.2 6 - 7 1024 125
GenBank mRNA AF218014.1 - 6 - 7 802 125
AK025899.1 - 6 - 7 954 125
AK025930.1 - 6 - 7 657 125
AK172846.1 - 6 - 7 959 125
AY562242.1 - 6 - 7 555 125
BC013048.1 - 6 - 7 1008 125
CR457353.1 - 6 - 7 555 125

View Full Product Details