Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R1
Entrez Gene ID
50834
Gene Name
taste 2 receptor member 1
Gene Aliases
T2R1, TRB7
Chromosome Location
Chr.5: 9628997 - 9630351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019599.2 NP_062545.1 1 - 1 1136 115
GenBank mRNA BC095521.1 - 1 - 1 1136 115
BC101729.1 - 1 - 1 890 115
BC101731.1 - 1 - 1 890 115

View Full Product Details