The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033557.2 | NP_001028729.1 | 1 - 2 | 467 | 105 |
NM_002706.5 | NP_002697.1 | 1 - 2 | 467 | 105 | |
NM_177968.3 | NP_808907.1 | 1 - 2 | 467 | 105 | |
XM_011532936.2 | XP_011531238.1 | 1 - 2 | 425 | 105 | |
XM_017004395.1 | XP_016859884.1 | 1 - 2 | 467 | 105 | |
XM_017004396.1 | XP_016859885.1 | 1 - 2 | 467 | 105 | |
XM_017004397.1 | XP_016859886.1 | 1 - 2 | 467 | 105 | |
GenBank mRNA | AK056009.1 | - | 1 - 2 | 403 | 105 |
AK291214.1 | - | 1 - 2 | 409 | 105 | |
AL833035.1 | - | 1 - 2 | 272 | 105 | |
BC064381.1 | - | 1 - 2 | 382 | 105 | |
BF979498.1 | - | 1 - 2 | 455 | 106 | |
DA128532.1 | - | 1 - 2 | 476 | 105 | |
DQ023508.1 | - | 1 - 2 | 393 | 105 | |
DQ023509.1 | - | 1 - 2 | 393 | 105 | |
DQ023510.1 | - | 1 - 2 | 393 | 105 | |
DQ023511.1 | - | 1 - 2 | 393 | 105 |