Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location
Chr.9: 110873252 - 111039259 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001401.3 NP_001392.2 3 - 4 1139 75
NM_057159.2 NP_476500.1 4 - 5 1217 75
XM_005251781.3 XP_005251838.1 4 - 5 1362 75
XM_005251782.3 XP_005251839.1 4 - 5 1456 75
XM_017014383.1 XP_016869872.1 4 - 5 1256 75
XM_017014384.1 XP_016869873.1 5 - 6 1630 75
XM_017014385.1 XP_016869874.1 6 - 7 1223 75
XM_017014386.1 XP_016869875.1 6 - 7 1523 75
XM_017014387.1 XP_016869876.1 6 - 7 1259 75
XM_017014388.1 XP_016869877.1 6 - 7 1442 75
XM_017014389.1 XP_016869878.1 5 - 6 1185 75
XM_017014390.1 XP_016869879.1 6 - 7 1220 75
XM_017014391.1 XP_016869880.1 6 - 7 1244 75
XM_017014392.1 XP_016869881.1 5 - 6 1487 75
XM_017014393.1 XP_016869882.1 6 - 7 1283 75
XM_017014394.1 XP_016869883.1 5 - 6 1412 75
XM_017014395.1 XP_016869884.1 5 - 6 1240 75
XM_017014396.1 XP_016869885.1 4 - 5 1182 75
XM_017014397.1 XP_016869886.1 5 - 6 1113 75
XM_017014398.1 XP_016869887.1 5 - 6 1356 75
XM_017014399.1 XP_016869888.1 5 - 6 1111 75
XM_017014400.1 XP_016869889.1 6 - 7 1472 75
XM_017014401.1 XP_016869890.1 5 - 6 1143 75
XM_017014402.1 XP_016869891.1 5 - 6 1200 75
XM_017014403.1 XP_016869892.1 6 - 7 1198 75
XM_017014404.1 XP_016869893.1 4 - 5 1396 75
XM_017014405.1 XP_016869894.1 6 - 7 1385 75
XM_017014406.1 XP_016869895.1 5 - 6 1119 75
XM_017014407.1 XP_016869896.1 5 - 6 1175 75
GenBank mRNA AK022808.1 - 3 - 4 1139 75
AK290287.1 - 5 - 6 1492 75
AK296374.1 - 2 - 3 820 75
AK298590.1 - 2 - 3 796 75
AK299279.1 - 2 - 3 870 75
AY322546.1 - 2 - 3 791 75
BC030615.2 - 4 - 5 1046 75
BC036034.1 - 4 - 5 1176 75
BC073167.1 - 5 - 6 1086 75
U78192.1 - 3 - 4 913 75
U80811.1 - 2 - 3 817 75
Y09479.1 - 3 - 4 913 75

View Full Product Details