Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD7
Entrez Gene ID
23424
Gene Name
tudor domain containing 7
Gene Aliases
CATC4, PCTAIRE2BP, TRAP
Chromosome Location
Chr.9: 97412020 - 97496125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302884.1 NP_001289813.1 11 - 12 2178 65
NM_014290.2 NP_055105.2 12 - 13 2391 65
XM_006717026.3 XP_006717089.1 11 - 12 1681 65
GenBank mRNA AB025254.1 - 3 - 4 276 65
AK294488.1 - 11 - 12 2104 65
AK314853.1 - 12 - 13 2303 65
BC028694.1 - 12 - 13 2390 65

View Full Product Details