Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD7
Entrez Gene ID
23424
Gene Name
tudor domain containing 7
Gene Aliases
CATC4, PCTAIRE2BP, TRAP
Chromosome Location
Chr.9: 97412020 - 97496125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302884.1 NP_001289813.1 8 - 9 1749 78
NM_014290.2 NP_055105.2 9 - 10 1962 78
XM_006717026.3 XP_006717089.1 8 - 9 1252 78
GenBank mRNA AK294488.1 - 8 - 9 1675 78
AK301954.1 - 2 - 3 330 78
AK314853.1 - 9 - 10 1874 78
BC028694.1 - 9 - 10 1961 78

View Full Product Details