The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206927.1 | NP_001193856.1 | 33 - 34 | 4574 | 69 |
XM_011514318.2 | XP_011512620.1 | 32 - 33 | 4522 | 69 | |
XM_011514319.2 | XP_011512621.1 | 33 - 34 | 4585 | 69 | |
XM_011514320.2 | XP_011512622.1 | 31 - 32 | 4348 | 69 | |
XM_011514321.1 | XP_011512623.1 | 30 - 31 | 3932 | 69 | |
XM_017010325.1 | XP_016865814.1 | 33 - 34 | 4585 | 69 | |
XM_017010326.1 | XP_016865815.1 | 33 - 34 | 4585 | 69 | |
XM_017010327.1 | XP_016865816.1 | 33 - 34 | 4586 | 69 | |
GenBank mRNA | AF356519.1 | - | 31 - 32 | 4068 | 69 |