The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175736.4 | NP_783863.4 | 11 - 12 | 1299 | 76 |
NM_198900.2 | NP_944489.2 | 10 - 11 | 1146 | 76 | |
XM_005269218.2 | XP_005269275.1 | 11 - 12 | 1278 | 76 | |
XM_011538968.2 | XP_011537270.1 | 11 - 12 | 1768 | 76 | |
XM_011538969.2 | XP_011537271.1 | 11 - 12 | 1767 | 76 | |
XM_011538971.2 | XP_011537273.1 | 11 - 12 | 1343 | 76 | |
XM_011538972.2 | XP_011537274.1 | 10 - 11 | 1613 | 76 | |
XM_011538973.2 | XP_011537275.2 | 7 - 8 | 758 | 76 | |
XM_011538974.2 | XP_011537276.1 | 12 - 13 | 1100 | 76 | |
GenBank mRNA | AB095934.2 | - | 11 - 12 | 1299 | 76 |
AK128195.1 | - | 10 - 11 | 1126 | 76 | |
BC159100.1 | - | 11 - 12 | 1299 | 76 |