The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012327.5 | NP_036459.1 | 21 - 22 | 2446 | 79 |
NM_176787.4 | NP_789744.1 | 22 - 23 | 2523 | 79 | |
XM_011525889.1 | XP_011524191.1 | 22 - 23 | 2491 | 79 | |
XM_011525890.1 | XP_011524192.1 | 22 - 23 | 2688 | 79 | |
XM_011525891.1 | XP_011524193.1 | 22 - 23 | 2406 | 79 | |
XM_011525892.1 | XP_011524194.1 | 21 - 22 | 2428 | 79 | |
XM_011525893.1 | XP_011524195.1 | 21 - 22 | 2379 | 79 | |
XM_011525894.1 | XP_011524196.1 | 21 - 22 | 2290 | 79 | |
XM_011525895.1 | XP_011524197.1 | 20 - 21 | 2288 | 79 | |
XM_011525896.1 | XP_011524198.1 | 22 - 23 | 2491 | 79 | |
XM_011525898.1 | XP_011524200.1 | 22 - 23 | 2491 | 79 | |
XM_017025685.1 | XP_016881174.1 | 19 - 20 | 2089 | 79 | |
XM_017025686.1 | XP_016881175.1 | 16 - 17 | 1679 | 79 | |
GenBank mRNA | AF109219.1 | - | 21 - 22 | 2359 | 79 |
AK000730.1 | - | 7 - 8 | 937 | 79 | |
AK315134.1 | - | 22 - 23 | 2491 | 79 | |
AL137607.1 | - | 18 - 19 | 1758 | 79 | |
BC028363.2 | - | 22 - 23 | 2460 | 79 |