The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032312.3 | NP_115688.1 | 1 - 2 | 353 | 135 |
XM_005264599.3 | XP_005264656.1 | 1 - 2 | 347 | 135 | |
XM_011533125.2 | XP_011531427.1 | 1 - 2 | 347 | 135 | |
GenBank mRNA | AB072898.1 | - | 1 - 2 | 300 | 135 |
AK098486.1 | - | 1 - 2 | 332 | 135 | |
BC004875.2 | - | 1 - 2 | 280 | 135 | |
BP207600.1 | - | 1 - 2 | 356 | 135 |