Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
Chromosome Location
Chr.16: 31214021 - 31225656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008274.3 NP_001008275.2 6 - 7 1141 71
XM_017023254.1 XP_016878743.1 5 - 6 677 71
GenBank mRNA AK090695.1 - 5 - 6 586 71
AK131485.1 - 6 - 7 1086 71

View Full Product Details