The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001895.2 | NP_001001895.1 | 4 - 5 | 638 | 67 |
NM_001243467.1 | NP_001230396.1 | 4 - 5 | 638 | 67 | |
GenBank mRNA | AF521702.1 | - | 4 - 5 | 565 | 67 |
AK308151.1 | - | 4 - 5 | 578 | 67 | |
AK313367.1 | - | 4 - 5 | 619 | 67 | |
BC028138.1 | - | 4 - 5 | 590 | 67 | |
BC069511.1 | - | 4 - 5 | 583 | 67 | |
BC069577.1 | - | 4 - 5 | 585 | 67 |