Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBASH3A
Entrez Gene ID
53347
Gene Name
ubiquitin associated and SH3 domain containing A
Gene Aliases
CLIP4, STS-2, TULA, TULA-1
Chromosome Location
Chr.21: 42403862 - 42447680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001895.2 NP_001001895.1 6 - 7 1013 73
NM_001243467.1 NP_001230396.1 6 - 7 1013 73
NM_018961.3 NP_061834.1 7 - 8 1127 73
XM_006724013.3 XP_006724076.1 7 - 8 1211 73
XM_011529605.2 XP_011527907.1 7 - 8 1127 73
XM_011529606.2 XP_011527908.1 7 - 8 1096 73
XM_011529607.2 XP_011527909.1 7 - 8 1180 73
XM_011529609.1 XP_011527911.1 7 - 8 1127 73
XM_011529610.2 XP_011527912.1 3 - 4 3126 73
GenBank mRNA AF520809.1 - 7 - 8 1054 73
AF521702.1 - 6 - 7 940 73
AJ277750.1 - 7 - 8 1074 73
AK308151.1 - 6 - 7 953 73
BC028138.1 - 6 - 7 965 73
BC069357.1 - 7 - 8 1074 73
BC069483.1 - 7 - 8 1074 73
BC069511.1 - 6 - 7 958 73
BC069577.1 - 6 - 7 960 73

View Full Product Details