Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
Chromosome Location
Chr.8: 94823290 - 94894664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017864.3 NP_060334.2 14 - 15 1892 133
NR_073444.1 14 - 15 1892 133
NR_073445.1 14 - 15 1861 133
XM_017013616.1 XP_016869105.1 14 - 15 1892 133
XM_017013617.1 XP_016869106.1 14 - 15 1892 133
XM_017013618.1 XP_016869107.1 11 - 12 1445 133
XM_017013619.1 XP_016869108.1 14 - 15 1851 133
GenBank mRNA AB161944.1 - 14 - 15 1876 133
AK000537.1 - 4 - 5 446 133
AK001403.1 - 7 - 8 832 133
AK091278.1 - 13 - 14 1629 133
AK225533.1 - 4 - 5 446 133
BC050536.1 - 6 - 7 680 133
BC064915.1 - 6 - 7 721 133
BC136754.1 - 14 - 15 1806 133
BC144396.1 - 14 - 15 1806 133
BC144397.1 - 14 - 15 1775 133
BX538203.1 - 10 - 11 1215 133

View Full Product Details