Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
Chromosome Location
Chr.8: 94823290 - 94894664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017864.3 NP_060334.2 9 - 10 1263 98
NR_073444.1 9 - 10 1263 98
NR_073445.1 9 - 10 1232 98
XM_017013616.1 XP_016869105.1 9 - 10 1263 98
XM_017013617.1 XP_016869106.1 9 - 10 1263 98
XM_017013618.1 XP_016869107.1 6 - 7 816 98
XM_017013619.1 XP_016869108.1 9 - 10 1222 98
GenBank mRNA AB161944.1 - 9 - 10 1247 98
AK001403.1 - 2 - 3 203 98
AK091278.1 - 8 - 9 1000 98
BC050536.1 - 1 - 2 51 98
BC064915.1 - 1 - 2 92 98
BC136754.1 - 9 - 10 1177 98
BC144396.1 - 9 - 10 1177 98
BC144397.1 - 9 - 10 1146 98
BX538203.1 - 5 - 6 586 98

View Full Product Details