The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193311.1 | NP_001180240.1 | 11 - 12 | 1029 | 79 |
NM_001193312.1 | NP_001180241.1 | 10 - 11 | 885 | 79 | |
NM_001193313.1 | NP_001180242.1 | 11 - 12 | 1029 | 79 | |
NM_024728.2 | NP_079004.1 | 11 - 12 | 918 | 79 | |
XM_006715775.3 | XP_006715838.1 | 11 - 12 | 1118 | 79 | |
XM_011515525.2 | XP_011513827.1 | 11 - 12 | 1025 | 79 | |
XM_011515526.2 | XP_011513828.1 | 11 - 12 | 1025 | 79 | |
XM_011515527.2 | XP_011513829.1 | 10 - 11 | 881 | 79 | |
XM_011515528.2 | XP_011513830.1 | 11 - 12 | 1025 | 79 | |
XM_011515529.2 | XP_011513831.1 | 10 - 11 | 881 | 79 | |
XM_017012621.1 | XP_016868110.1 | 11 - 12 | 1134 | 79 | |
XM_017012622.1 | XP_016868111.1 | 11 - 12 | 1025 | 79 | |
GenBank mRNA | AB014767.1 | - | 10 - 11 | 882 | 79 |
AK021870.1 | - | 11 - 12 | 918 | 79 | |
AK296048.1 | - | 11 - 12 | 996 | 79 | |
AK299133.1 | - | 11 - 12 | 1009 | 79 | |
AK307935.1 | - | 11 - 12 | 988 | 79 | |
AK308764.1 | - | 11 - 12 | 989 | 79 | |
BC098117.1 | - | 11 - 12 | 1028 | 79 | |
BC098261.1 | - | 11 - 12 | 1028 | 79 | |
BC098310.1 | - | 10 - 11 | 884 | 79 | |
BC098318.1 | - | 11 - 12 | 1028 | 79 |