The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193311.1 | NP_001180240.1 | 14 - 15 | 1279 | 90 |
NM_001193312.1 | NP_001180241.1 | 12 - 13 | 1057 | 90 | |
NM_001193313.1 | NP_001180242.1 | 13 - 14 | 1201 | 90 | |
NM_024728.2 | NP_079004.1 | 14 - 15 | 1168 | 90 | |
XM_006715775.3 | XP_006715838.1 | 14 - 15 | 1368 | 90 | |
XM_011515525.2 | XP_011513827.1 | 15 - 16 | 1353 | 90 | |
XM_011515526.2 | XP_011513828.1 | 15 - 16 | 1353 | 90 | |
XM_011515527.2 | XP_011513829.1 | 14 - 15 | 1209 | 90 | |
XM_011515529.2 | XP_011513831.1 | 13 - 14 | 1131 | 90 | |
XM_017012621.1 | XP_016868110.1 | 15 - 16 | 1462 | 90 | |
GenBank mRNA | AB014767.1 | - | 12 - 13 | 1054 | 90 |
AK021870.1 | - | 14 - 15 | 1168 | 90 | |
AK296048.1 | - | 13 - 14 | 1168 | 90 | |
AK299133.1 | - | 13 - 14 | 1181 | 90 | |
AK307935.1 | - | 13 - 14 | 1160 | 90 | |
BC098117.1 | - | 13 - 14 | 1200 | 90 | |
BC098261.1 | - | 13 - 14 | 1200 | 90 | |
BC098310.1 | - | 12 - 13 | 1056 | 90 | |
BC098318.1 | - | 14 - 15 | 1278 | 90 | |
BM668297.1 | - | 1 - 2 | 90 |