The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193311.1 | NP_001180240.1 | 3 - 4 | 267 | 96 |
NM_001193312.1 | NP_001180241.1 | 3 - 4 | 267 | 96 | |
NM_001193313.1 | NP_001180242.1 | 3 - 4 | 267 | 96 | |
NM_024728.2 | NP_079004.1 | 3 - 4 | 267 | 96 | |
XM_006715775.3 | XP_006715838.1 | 3 - 4 | 356 | 96 | |
XM_011515525.2 | XP_011513827.1 | 3 - 4 | 263 | 96 | |
XM_011515526.2 | XP_011513828.1 | 3 - 4 | 263 | 96 | |
XM_011515527.2 | XP_011513829.1 | 3 - 4 | 263 | 96 | |
XM_011515528.2 | XP_011513830.1 | 3 - 4 | 263 | 96 | |
XM_011515529.2 | XP_011513831.1 | 3 - 4 | 263 | 96 | |
XM_011515530.2 | XP_011513832.1 | 3 - 4 | 263 | 96 | |
XM_017012621.1 | XP_016868110.1 | 3 - 4 | 372 | 96 | |
XM_017012622.1 | XP_016868111.1 | 3 - 4 | 263 | 96 | |
GenBank mRNA | AB014767.1 | - | 2 - 3 | 120 | 96 |
AK021870.1 | - | 3 - 4 | 267 | 96 | |
AK296048.1 | - | 3 - 4 | 234 | 96 | |
AK298516.1 | - | 3 - 4 | 223 | 96 | |
AK299133.1 | - | 3 - 4 | 247 | 96 | |
AK307935.1 | - | 3 - 4 | 225 | 96 | |
AK308764.1 | - | 3 - 4 | 227 | 96 | |
BC098117.1 | - | 3 - 4 | 266 | 96 | |
BC098261.1 | - | 3 - 4 | 266 | 96 | |
BC098310.1 | - | 3 - 4 | 266 | 96 | |
BC098318.1 | - | 3 - 4 | 266 | 96 |