The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042599.1 | NP_001036064.1 | 4 - 5 | 647 | 75 |
NM_005235.2 | NP_005226.1 | 4 - 5 | 647 | 75 | |
XM_005246376.2 | XP_005246433.1 | 4 - 5 | 1166 | 75 | |
XM_005246377.2 | XP_005246434.1 | 4 - 5 | 1166 | 75 | |
XM_006712364.2 | XP_006712427.1 | 4 - 5 | 1166 | 75 | |
GenBank mRNA | AK308606.1 | - | 4 - 5 | 603 | 75 |
BC112199.1 | - | 4 - 5 | 588 | 75 | |
BC143741.1 | - | 4 - 5 | 576 | 75 | |
BC143747.1 | - | 4 - 5 | 576 | 75 | |
BC143749.1 | - | 4 - 5 | 576 | 75 | |
L07868.1 | - | 4 - 5 | 582 | 75 |